Ethyl 5-oxopyrrolidine-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: N-DL-5-oxo-Pro-OEt

A861035-100G A861035-25G A861035-5G A861035-1G
77105-970EA 203.7 USD
77105-970 77105-972 77105-974 77614-546
Ethyl 5-oxopyrrolidine-2-carboxylate 98%
Ethyl 5-oxopyrrolidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃
MW: 157.16 g/mol
MDL Number: MFCD00062673
CAS Number: 66183-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR