3-(Chloromethyl)-5-methoxypyridine hydrochloride 95%

Supplier: Ambeed
Danger

A856108-100MG A856108-250MG
77105-952EA 106.09 USD
77105-952 77105-954
3-(Chloromethyl)-5-methoxypyridine hydrochloride 95%
3-(Chloromethyl)-5-methoxypyridine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉Cl₂NO
MW: 194.05 g/mol
MDL Number: MFCD28506091
CAS Number: 1801422-13-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR