(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(2-(tert-butoxy)-2-oxoethyl)phenyl)propanoic acid 98%

Supplier: Ambeed
Warning

A855891-100MG A855891-1G A855891-250MG
77105-950EA 120.95 USD
77105-950 77569-586 77595-376
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(2-(tert-butoxy)-2-oxoethyl)phenyl)propanoic acid 98%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-(2-(tert-butoxy)-2-oxoethyl)phenyl)propanoic acid

Precaución: For research use only.

Formula: C₃₀H₃₁NO₆
MW: 501.57 g/mol
MDL Number: MFCD14586316
CAS Number: 222842-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR