2-Ethylmorpholine hydrochloride 98%

Supplier: Ambeed
Warning

A847888-100MG A847888-1G A847888-250MG A847888-5G
77105-914EA 7 USD
77105-914 77105-916 77105-918 77901-949
2-Ethylmorpholine hydrochloride 98%
2-Ethylmorpholine hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄ClNO
MDL Number: MFCD14705612
CAS Number: 1221722-35-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR