DL-Histidine monohydrochloride monohydrate 99+%

Supplier: Ambeed
Warning

Synonyms: H-DL-His-OH.HCl.H2O, H-DL-His-OH·HCl·H₂O

A847191-100G A847191-10G A847191-1G A847191-25G A847191-5G
77105-888EA 120.95 USD
77105-888 77105-890 77105-892 77105-894 77105-896
DL-Histidine monohydrochloride monohydrate 99+%
DL-Histidine monohydrochloride monohydrate

Precaución: for Research Use Only

Formula: C₆H₉N₃O₂·HCl·H₂O
MW: 209.63 g/mol (191.62 anhy)
MDL Number: MFCD00151028
CAS Number: 123333-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR