2-(3,4-Dichlorophenyl)-2-methylpropanoic acid 98%

Supplier: Ambeed
Warning

A845187-100MG A845187-250MG
77105-884EA 33 USD
77105-884 77105-886
2-(3,4-Dichlorophenyl)-2-methylpropanoic acid 98%
2-(3,4-Dichlorophenyl)-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₂
MDL Number: MFCD11036926
CAS Number: 80854-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR