2,4,6-Trifluorobenzyl bromide 97%

Supplier: Ambeed
Danger

A838192-1G A838192-5G A838192-25G A838192-250MG
77105-858EA 37 USD
77105-858 77105-860 77686-868 77900-734
2,4,6-Trifluorobenzyl bromide 97%
2,4,6-Trifluorobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₄BrF₃
MDL Number: MFCD00061210
CAS Number: 151411-98-2
UN: 2922
ADR: 8,6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR