1,3-Dibromo-7-tert-butylpyrene 98%

Supplier: Ambeed
Warning

A836117-1G A836117-250MG A836117-5G
77105-848EA 83 USD
77105-848 77105-850 77105-852
1,3-Dibromo-7-tert-butylpyrene 98%
1,3-Dibromo-7-tert-butylpyrene

Precaución: for Research Use Only

Formula: C₂₀H₁₆Br₂
MDL Number: MFCD22581440
CAS Number: 1005771-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR