(6-Amino-2,3-difluorophenyl)acetic acid 95%

Supplier: Ambeed
Warning

A835368-1G A835368-250MG
77105-844EA 246 USD
77105-844 77105-846
(6-Amino-2,3-difluorophenyl)acetic acid 95%
(6-Amino-2,3-difluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₇F₂NO₂
MDL Number: MFCD07368097
CAS Number: 887585-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR