(S)-Methyl-2-aminohexanoate hydrochloride 98%

Supplier: Ambeed
Warning

A835017-1G A835017-25G A835017-5G
77105-836EA 6 USD
77105-836 77105-838 77105-840
(S)-Methyl-2-aminohexanoate hydrochloride 98%
(S)-Methyl-2-aminohexanoate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₆ClNO₂
MDL Number: MFCD00077151
CAS Number: 3844-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR