tert-Butyl 3-cyanobenzoate 97%

Supplier: Ambeed
Warning

A834428-100MG A834428-1G
77105-830EA 6 USD
77105-830 77105-832
tert-Butyl 3-cyanobenzoate 97%
tert-Butyl 3-cyanobenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₂
MDL Number: MFCD16620851
CAS Number: 383185-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR