Z-Gly-OSu 98%

Supplier: Ambeed
Warning

Synonyms: Z-Gly-Osu

A833814-1G A833814-5G A833814-25G
77105-826EA 11.67 USD
77105-826 77105-828 77614-498
Z-Gly-OSu 98%
Z-Gly-OSu

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂O₆
MW: 306.274 g/mol
MDL Number: MFCD00153332
CAS Number: 2899-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR