(2,4,5-Trifluorophenyl)acetonitrile 98%

Supplier: Ambeed
Danger

A833364-1G A833364-5G A833364-25G
77105-820EA 11 USD
77105-820 77105-822 77105-824 77686-860
(2,4,5-Trifluorophenyl)acetonitrile 98%
(2,4,5-Trifluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₄F₃N
MDL Number: MFCD00061220
CAS Number: 220141-74-2
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR