3,4-(Methylenedioxy)benzylamine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Piperonylamine, Benzo[d][1,3]dioxol-5-ylmethanamine
A833293-100G
A833293-10G
A833293-25G
A833293-5G
77105-812EA
259
USD
77105-812
77105-814
77105-816
77105-818
3,4-(Methylenedioxy)benzylamine 95%
3,4-(Methylenedioxy)benzylamine
Precaución: for Research Use Only
Formula:
C₈H₉NO₂ |
MDL Number:
MFCD00005840 CAS Number: 2620-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...