2,3-Difluoro-4-iodoaniline 95%

Supplier: Ambeed
Warning

A833229-1G A833229-250MG
77105-808EA 31 USD
77105-808 77105-810
2,3-Difluoro-4-iodoaniline 95%
2,3-Difluoro-4-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄F₂IN
MDL Number: MFCD21602052
CAS Number: 1026989-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR