2,3-Dichloro-6-(trifluoromethyl)benzyl bromide 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(Bromomethyl)-3,4-dichloro-1-(trifluoromethyl)benzene
A832449-1G
A832449-250MG
77105-804EA
62.6
USD
77105-804
77105-806
2,3-Dichloro-6-(trifluoromethyl)benzyl bromide 95%
2,3-Dichloro-6-(trifluoromethyl)benzyl bromide
Precaución: for Research Use Only
Formula:
C₈H₄BrCl₂F₃ MW: 307.93 g/mol |
MDL Number:
MFCD06660275 CAS Number: 886501-99-1 UN: 3265 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...