Ethyl 4,5-Difluoro-2-nitrobenzoate 95%

Supplier: Ambeed
Warning

A831670-10G A831670-1G A831670-5G
77105-798EA 129.43 USD
77105-798 77105-800 77105-802
Ethyl 4,5-Difluoro-2-nitrobenzoate 95%
Ethyl 4,5-Difluoro-2-nitrobenzoate

Precaución: for Research Use Only

Formula: C₉H₇F₂NO₄
MW: 231.153 g/mol
MDL Number: MFCD11042826
CAS Number: 1214387-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR