2-Bromo-7-methoxynaphthalene 95%

Supplier: Ambeed
Warning

A829899-100MG A829899-250MG A829899-1G A829899-5G
77105-794EA 61.54 USD
77105-794 77105-796 77595-322 77686-854
2-Bromo-7-methoxynaphthalene 95%
2-Bromo-7-methoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₁H₉BrO
MW: 237.093 g/mol
MDL Number: MFCD17012538
CAS Number: 200875-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR