Sodium 3,5-difluorobenzoate 95+%

Supplier: Ambeed
Warning

A828046-100G A828046-25G A828046-5G
77105-788EA 86 USD
77105-788 77105-790 77105-792
Sodium 3,5-difluorobenzoate 95+%
Sodium 3,5-difluorobenzoate

Precaución: for Research Use Only

Formula: C₇H₃F₂NaO₂
MDL Number: MFCD03788553
CAS Number: 530141-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR