(S)-5-Benzyl-5-azaspiro[2.4]heptan-7-amine 98%

Supplier: Ambeed
Warning

A827644-100MG A827644-1G A827644-250MG
77105-782EA 51.99 USD
77105-782 77105-784 77105-786
(S)-5-Benzyl-5-azaspiro[2.4]heptan-7-amine 98%
(S)-5-Benzyl-5-azaspiro[2.4]heptan-7-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₈N₂
MW: 202.29 g/mol
MDL Number: MFCD17926316
CAS Number: 144282-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR