(S)-Di-tert-butyl 2-(3-((S)-6-amino-1-tert-butoxy-1-oxohexan-2-yl)ureido)pentanedioate 95%

Supplier: Ambeed
Warning

Synonyms: CS-0047390

A827631-100MG A827631-1G A827631-250MG A827631-5G
77105-780EA 167.63 USD
77105-780 77552-534 77552-536 77686-853
(S)-Di-tert-butyl 2-(3-((S)-6-amino-1-tert-butoxy-1-oxohexan-2-yl)ureido)pentanedioate 95%
(S)-Di-tert-butyl 2-(3-((S)-6-amino-1-tert-butoxy-1-oxohexan-2-yl)ureido)pentanedioate

Precaución: for Research Use Only

Formula: C₂₄H₄₅N₃O₇
MW: 487.63 g/mol
MDL Number: MFCD28898592
CAS Number: 1025796-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR