5-Chloro-2,1,3-benzothiadiazol-4-ylamine 98%

Supplier: Ambeed
Warning

A827269-100G A827269-1G A827269-25G A827269-500G A827269-5G
77105-770EA 265.23 USD
77105-770 77105-772 77105-774 77105-776 77105-778
5-Chloro-2,1,3-benzothiadiazol-4-ylamine 98%
5-Chloro-2,1,3-benzothiadiazol-4-ylamine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃S
MW: 185.6341 g/mol
MDL Number: MFCD00465908
CAS Number: 30536-19-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR