3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol 90%

Supplier: Ambeed
Warning

A826694-100MG A826694-1G A826694-250MG
77105-764EA 52 USD
77105-764 77105-766 77105-768
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol 90%
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol

Precaución: for Research Use Only

Formula: C₁₆H₁₉BO₃
MDL Number: MFCD19237213
CAS Number: 1437769-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR