2-(2-Bromophenyl)propan-2-amine hydrochloride 95%

Supplier: Ambeed
Warning

A821853-100MG A821853-250MG
77105-754EA 54.11 USD
77105-754 77105-756
2-(2-Bromophenyl)propan-2-amine hydrochloride 95%
2-(2-Bromophenyl)propan-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃BrClN
MW: 250.56 g/mol
MDL Number: MFCD11870089
CAS Number: 1087723-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR