(S)-3-(1-Amino-3-hydroxypropyl)phenol hydrochloride 95%

Supplier: Ambeed
Warning

A820945-100MG A820945-1G A820945-250MG
77105-748EA 53.05 USD
77105-748 77105-750 77105-752
(S)-3-(1-Amino-3-hydroxypropyl)phenol hydrochloride 95%
(S)-3-(1-Amino-3-hydroxypropyl)phenol hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄ClNO₂
MW: 203.66 g/mol
MDL Number: MFCD11053647
CAS Number: 1212827-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR