Potassium trifluoro(1,4-dioxaspiro[4.5]dec-7-en-8-yl)borate 98%

Supplier: Ambeed
Danger

A820412-1G A820412-250MG A820412-5G
77105-740EA 201.58 USD
77105-740 77105-742 77105-744
Potassium trifluoro(1,4-dioxaspiro[4.5]dec-7-en-8-yl)borate 98%
Potassium trifluoro(1,4-dioxaspiro[4.5]dec-7-en-8-yl)borate

Precaución: for Research Use Only

Formula: C₈H₁₁BF₃KO₂
MW: 246.08 g/mol
MDL Number: MFCD09993678
CAS Number: 1580472-97-4
UN: 1759
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR