4-Chloro-3,5-dibromotoluene 95%

Supplier: Ambeed
Warning

A820216-100G A820216-10G A820216-1G A820216-5G
77105-722EA 392.54 USD
77105-722 77105-724 77105-726 77105-728
4-Chloro-3,5-dibromotoluene 95%
4-Chloro-3,5-dibromotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂Cl
MW: 284.37 g/mol
MDL Number: MFCD01861197
CAS Number: 202925-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR