1,3-Diisopropylthiourea 99%
Supplier: Ambeed
|
Warning
|
A818641-1000G
A818641-100G
A818641-500G
A818641-25G
77105-712EA
116
USD
77105-712
77105-714
77105-716
77308-378
1,3-Diisopropylthiourea 99%
1,3-Diisopropylthiourea
Precaución: for Research Use Only
Formula:
C₇H₁₆N₂S |
MDL Number:
MFCD00040485 CAS Number: 2986-17-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...