tert-Butyl 1-oxo-1,3-dihydrospiro[indene-2,4'-piperidine]-1'-carboxylate 98%

Supplier: Ambeed
Warning

A816748-100MG A816748-1G A816748-250MG A816748-5G
77105-694EA 50.93 USD
77105-694 77105-696 77105-698 77105-700
tert-Butyl 1-oxo-1,3-dihydrospiro[indene-2,4'-piperidine]-1'-carboxylate 98%
tert-Butyl 1-oxo-1,3-dihydrospiro[indene-2,4'-piperidine]-1'-carboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₃NO₃
MW: 301.38 g/mol
MDL Number: MFCD22689932
CAS Number: 1228079-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR