2-(3-Chloro-4-isopropoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-4-isopropoxyphenylboronic acid pinacol ester

A816144-1G A816144-250MG A816144-5G
77105-688EA 94.43 USD
77105-688 77105-690 77105-692
2-(3-Chloro-4-isopropoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(3-Chloro-4-isopropoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₂₂BClO₃
MW: 296.60 g/mol
MDL Number: MFCD18384181
CAS Number: 1260023-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR