(1aS,1bS,2S,5aR,6S,6aS)-1a-(Hydroxymethyl)-2-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1a,1b,2,5a,6,6a-hexahydrooxireno[2',3':4,5]cyclopenta[1,2-c]pyran-6-yl 4-hydroxy-3-methoxybenzoate 95%

Supplier: Ambeed
Warning

A815479-10MG A815479-1MG A815479-25MG A815479-50MG
77105-680EA 38 USD
77105-680 77105-682 77105-684 77105-686
(1aS,1bS,2S,5aR,6S,6aS)-1a-(Hydroxymethyl)-2-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1a,1b,2,5a,6,6a-hexahydrooxireno[2',3':4,5]cyclopenta[1,2-c]pyran-6-yl 4-hydroxy-3-methoxybenzoate 95%
(1aS,1bS,2S,5aR,6S,6aS)-1a-(Hydroxymethyl)-2-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-1a,1b,2,5a,6,6a-hexahydrooxireno[2',3':4,5]cyclopenta[1,2-c]pyran-6-yl 4-hydroxy-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₂₃H₂₈O₁₃
MDL Number: MFCD00135962
CAS Number: 39012-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR