Methyl 3-bromo-5-chloro-2-methylbenzoate 95%

Supplier: Ambeed
Warning

A814709-100MG A814709-250MG A814709-1G
77105-676EA 50 USD
77105-676 77105-678 77552-506
Methyl 3-bromo-5-chloro-2-methylbenzoate 95%
Methyl 3-bromo-5-chloro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
MDL Number: MFCD24108598
CAS Number: 1403597-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR