ELR-510444 98+%

Supplier: Ambeed
Warning

Synonyms: N-(5-(5-Cyanothiophen-2-yl)-2-methylphenyl)-4-methylbenzenesulphonamide, ELR510444

A814378-10MG A814378-1MG A814378-50MG A814378-5MG
77105-668EA 33.95 USD
77105-668 77105-670 77105-672 77105-674
ELR-510444 98+%
ELR-510444

Precaución: for Research Use Only

Formula: C₁₉H₁₆N₂O₂S₂
MW: 368.5 g/mol
MDL Number: MFCD25976813
CAS Number: 1233948-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR