4-(2,2,2-Trifluoroethoxy)phenylhydrazine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 1-Hydrazino-4-(2,2,2-trifluoroethoxy)benzene hydrochloride, 4-Hydrazino-β,β,β-trifluorophenetole hydrochloride, (4-(2,2,2-Trifluoroethoxy)phenyl)hydrazine hydrochloride

A812948-100MG A812948-1G A812948-250MG
77105-658EA 62 USD
77105-658 77105-660 77105-662
4-(2,2,2-Trifluoroethoxy)phenylhydrazine hydrochloride 98%
4-(2,2,2-Trifluoroethoxy)phenylhydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉F₃N₂O·ClH
MDL Number: MFCD09763680
CAS Number: 115171-04-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR