2-Bromo-5-(methanesulfonyl)pyridine 97%

Supplier: Ambeed
Warning

A811281-1G A811281-250MG A811281-5G
77105-652EA 36 USD
77105-652 77105-654 77105-656
2-Bromo-5-(methanesulfonyl)pyridine 97%
2-Bromo-5-(methanesulfonyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
MDL Number: MFCD04974509
CAS Number: 343262-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR