1,3-Dimethoxy-4-iodobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Iodo-2,4-dimethoxybenzene, 2,4-Dimethoxyiodobenzene

A810874-1G A810874-250MG A810874-25G A810874-5G
77105-644EA 11.67 USD
77105-644 77105-646 77105-648 77105-650
1,3-Dimethoxy-4-iodobenzene 98%
1,3-Dimethoxy-4-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₉IO₂
MW: 264.06 g/mol
MDL Number: MFCD00059268
CAS Number: 20469-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR