5-Hydroxy-2-(trifluoromethoxy)phenylboronic acid 98%

Supplier: Ambeed
Warning

A806915-1G A806915-250MG
77105-628EA 114.58 USD
77105-628 77105-630
5-Hydroxy-2-(trifluoromethoxy)phenylboronic acid 98%
5-Hydroxy-2-(trifluoromethoxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BF₃O₄
MW: 221.926 g/mol
MDL Number: MFCD12913937
CAS Number: 1217500-69-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR