2-(4-Methoxy-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%

Supplier: Ambeed
Warning

A806757-1G A806757-250MG A806757-5G
77105-622EA 51.99 USD
77105-622 77105-624 77105-626
2-(4-Methoxy-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-(4-Methoxy-3,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₅H₂₃BO₃
MW: 262.15 g/mol
MDL Number: MFCD22414439
CAS Number: 568572-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR