3-Iodo-4-methoxybenzonitrile 95%

Supplier: Ambeed
Warning

A805488-1G A805488-250MG
77105-612EA 123 USD
77105-612 77901-161
3-Iodo-4-methoxybenzonitrile 95%
3-Iodo-4-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆INO
MDL Number: MFCD17010713
CAS Number: 82504-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR