2-Hydroxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 95%

Supplier: Ambeed
Warning

A803092-100MG A803092-1G A803092-250MG
77105-606EA 44.56 USD
77105-606 77105-608 77105-610
2-Hydroxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 95%
2-Hydroxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₃
MW: 245.08 g/mol
MDL Number: MFCD16994359
CAS Number: 1350933-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR