2-Amino-4-chloro-5-methoxybenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A802669-100MG
A802669-250MG
A802669-5G
A802669-1G
77105-600EA
29.71
USD
77105-600
77105-602
77105-604
77614-430
2-Amino-4-chloro-5-methoxybenzoic acid 95%
2-Amino-4-chloro-5-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₈ClNO₃ MW: 201.60 g/mol |
MDL Number:
MFCD22568950 CAS Number: 181434-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...