2,6-Di(1-pyrazolyl)pyridine 98%

Supplier: Ambeed
Warning

A802070-100MG A802070-1G A802070-250MG A802070-5G A802070-25G
77105-592EA 9 USD
77105-592 77105-594 77105-596 77105-598 77901-374
2,6-Di(1-pyrazolyl)pyridine 98%
2,6-Di(1-pyrazolyl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₉N₅
MDL Number: MFCD28336856
CAS Number: 123640-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR