2,4-Difluorobenzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-2,4-difluorobenzene, α-Bromo-2,4-difluorotoluene

A802027-5G A802027-25G A802027-10G
77105-590EA 12 USD
77105-590 77552-468 77686-823
2,4-Difluorobenzyl bromide 97%
2,4-Difluorobenzyl bromide

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MDL Number: MFCD00011649
CAS Number: 23915-07-3
UN: 2920
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR