6-Chloro-2-aminobenzamide 97%

Supplier: Ambeed
Warning

A801692-100MG A801692-250MG
77105-586EA 37 USD
77105-586 77105-588
6-Chloro-2-aminobenzamide 97%
6-Chloro-2-aminobenzamide

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O
MDL Number: MFCD00234296
CAS Number: 54166-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR