(S)-4-Benzyl-3-pentanoyloxazolidin-2-one 98%

Supplier: Ambeed
Warning

A797906-100MG A797906-1G A797906-250MG
77105-578EA 61 USD
77105-578 77105-580 77105-582
(S)-4-Benzyl-3-pentanoyloxazolidin-2-one 98%
(S)-4-Benzyl-3-pentanoyloxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₃
MDL Number: MFCD20269711
CAS Number: 143868-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR