Rosmarinic acid 98%

Supplier: Ambeed
Warning

A796919-100MG A796919-10MG A796919-1G A796919-1MG A796919-250MG A796919-25G A796919-5G
77105-564EA 13 USD
77105-564 77105-566 77105-568 77105-570 77105-572 77105-574 77105-576
Rosmarinic acid 98%
Rosmarinic acid

Precaución: for Research Use Only

Formula: C₁₈H₁₆O₈
MDL Number: MFCD00017740
CAS Number: 20283-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR