2-Fluoro-5-nitrobenzoyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 3-(Chloroformyl)-4-fluoronitrobenzene, 3-(Chlorocarbonyl)-4-fluoronitrobenzene

A796514-1G A796514-5G A796514-25G
77105-560EA 13.8 USD
77105-560 77105-562 77686-821
2-Fluoro-5-nitrobenzoyl chloride 98%
2-Fluoro-5-nitrobenzoyl chloride

Precaución: for Research Use Only

Formula: C₇H₃ClFNO₃
MW: 203.555 g/mol
MDL Number: MFCD08458077
CAS Number: 709-46-6
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR