4-Ethyl-2,6-difluorophenol 95%

Supplier: Ambeed
Warning

A795176-1G A795176-250MG
77105-552EA 198.39 USD
77105-552 77105-554
4-Ethyl-2,6-difluorophenol 95%
4-Ethyl-2,6-difluorophenol

Precaución: for Research Use Only

Formula: C₈H₈F₂O
MW: 158.14 g/mol
MDL Number: MFCD28163189
CAS Number: 326493-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR