3-Methoxybenzylamine 98%

Supplier: Ambeed
Danger

Synonyms: m-Anisylamine, 3-(Aminomethyl)anisole

A795160-10G A795160-1G A795160-5G
77105-546EA 13 USD
77105-546 77105-548 77105-550
3-Methoxybenzylamine 98%
3-Methoxybenzylamine

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MDL Number: MFCD00008115
CAS Number: 5071-96-5
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR