2,4,5-Triiodoimidazole 98%

Supplier: Ambeed
Danger

A795120-10G A795120-1G A795120-250MG A795120-5G
77105-538EA 53 USD
77105-538 77105-540 77105-542 77105-544
2,4,5-Triiodoimidazole 98%
2,4,5-Triiodoimidazole

Precaución: for Research Use Only

Formula: C₃HI₃N₂
MDL Number: MFCD00068812
CAS Number: 1746-25-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR